(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 98%

Warning

Synonyms: H-4-Cyano-D-Phe-OH

H63687-03 H63687-MD H63687-06
AAH63687-03EA 128.76 USD
AAH63687-03 AAH63687-06 AAH63687-MD
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 98%
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid
Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270364
CAS Number: 263396-44-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR