1-Methyl-4-(4-nitrophenyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H63665-03 H63665-MD H63665-06
AAH63665-06EA 231.99 USD
AAH63665-06 AAH63665-03 AAH63665-MD
1-Methyl-4-(4-nitrophenyl)piperazine 97%
1-Methyl-4-(4-nitrophenyl)piperazine
Formula: C11H15N3O2
MW: 221.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00101003
CAS Number: 16155-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR