1-Ethyl-4-(4-nitrophenyl)piperazine 97%

Warning

H63555-03 H63555-06 H63555-MD
AAH63555-03EA 66.44 USD
AAH63555-03 AAH63555-06 AAH63555-MD
1-Ethyl-4-(4-nitrophenyl)piperazine 97%
1-Ethyl-4-(4-nitrophenyl)piperazine
Formula: C12H17N3O2
Storage Temperature: Ambient
MDL Number: MFCD00614372
CAS Number: 115619-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR