4-(2-Fluoro-6-nitrophenyl)morpholine 97%

Supplier: Thermo Scientific Chemicals
Warning

H63272-MD H63272-03 H63272-06
AAH63272-MDEA 23.23 USD
AAH63272-MD AAH63272-03 AAH63272-06
4-(2-Fluoro-6-nitrophenyl)morpholine 97%
4-(2-Fluoro-6-nitrophenyl)morpholine
Formula: C₁₀H₁₁FN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD13689260
CAS Number: 1233955-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR