3-Fluoro-4-methylphenylboronic acid pinacol ester ≥96%

Synonyms: 2-(3-Fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane3-Fluoro-4-methylbenzeneboronic acid pinacol ester

H62872-03 H62872-MD
AAH62872-03EA 278.81 USD
AAH62872-03 AAH62872-MD
3-Fluoro-4-methylphenylboronic acid pinacol ester ≥96%
3-Fluoro-4-methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₈BFO₂
MW: 236.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972178
CAS Number: 903895-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR