2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-(4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate2-Fluoro-5-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester

H62786-03 H62786-MD
AAH62786-MDEA 168.99 USD
AAH62786-MD AAH62786-03
2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester 96%
2-Fluoro-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester
Formula: C₁₅H₂₀BFO₄
MW: 294.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026092
CAS Number: 944317-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR