5-Cyano-2-fluorobenzeneboronic acid pinacol ester 96%

Warning

Synonyms: 4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile5-Cyano-2-fluorophenylboronic acid pinacol ester

H62739-03 H62739-MD
AAH62739-03EA 381.15 USD
AAH62739-03 AAH62739-MD
5-Cyano-2-fluorobenzeneboronic acid pinacol ester 96%
5-Cyano-2-fluorobenzeneboronic acid pinacol ester
Formula: C₁₃H₁₅BFNO₂
MW: 247.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795681
CAS Number: 775351-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR