2-Cyano-4-fluorophenylboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyano-4-fluorophenylboronic Acid Pinacol Ester, 2-Cyano-4-fluorobenzeneboronic acid pinacol ester

H62729-MD
AAH62729-03EA 289.71 USD
AAH62729-03 AAH62729-MD
2-Cyano-4-fluorophenylboronic acid pinacol ester 96%
2-Cyano-4-fluorophenylboronic acid pinacol ester
Formula: C₁₃H₁₅BFNO₂
MW: 247.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09260444
CAS Number: 461451-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR