2-Fluoro-5-formylphenylboronic acid pinacol ester ≥96%

Synonyms: 2-Fluoro-5-formylphenylboronic acid, pinacol ester2-Fluoro-5-formylbenzeneboronic acid pinacol ester4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

H62689-MD H62689-06
AAH62689-06EA 558.69 USD
AAH62689-06 AAH62689-MD
2-Fluoro-5-formylphenylboronic acid pinacol ester ≥96%
2-Fluoro-5-formylphenylboronic acid pinacol ester
Formula: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
Melting Pt: 35…40 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD11867877
CAS Number: 1112208-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR