2-Benzyloxy-4-fluorophenylboronic acid pinacol ester ≥96%

Synonyms: 2-Benzyloxy-4-fluorobenzeneboronic acid pinacol ester

H62541-03 H62541-MD
AAH62541-03EA 485.74 USD
AAH62541-03 AAH62541-MD
2-Benzyloxy-4-fluorophenylboronic acid pinacol ester ≥96%
2-Benzyloxy-4-fluorophenylboronic acid pinacol ester
Formula: C₁₉H₂₂BFO₃
MW: 246.046 g/mol
Storage Temperature: Ambient
CAS Number: 848779-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR