2-Bromo-2',4'-difluoroacetophenone 97%

Danger

Synonyms: 2,4-Difluorophenacyl bromide2-Bromo-1-(2,4-difluorophenyl)ethanone

H56442-03 H56442-06 H56442-MD
AAH56442-03EA 83.77 USD
AAH56442-03 AAH56442-MD AAH56442-06
2-Bromo-2',4'-difluoroacetophenone 97%
2-Bromo-2',4'-difluoroacetophenone
Formula: C₈H₅BrF₂O
MW: 235.03 g/mol
Melting Pt: 31…35 °C
Flash Pt: 51 °C(124 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142822
CAS Number: 102429-07-2
UN: 2925
ADR: "4.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR