1-Ethylnaphthalene 98%

Supplier: Thermo Scientific Chemicals

H56413-14 H56413-06
AAH56413-14EA 467.99 USD
AAH56413-14 AAH56413-06
1-Ethylnaphthalene 98%
1-Ethylnaphthalene
Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 258…260 °C
Melting Pt: –15 °C
Density: 1.008
Flash Pt: 111 °C(232 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004049
CAS Number: 1127-76-0
EINECS: 214-432-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR