(2S)-(+)-2,3-Epoxypropyl toluene-4-sulfonate 99%

Danger

Synonyms: (2S)-(+)-Glycidyl tosylate(s)-(+)-oxirane-2-methanol p-toluenesulfonate(2S)-(+)-Glycidyl p-toluenesulfonate(2S)-(+)-Glycidyl p-toluenesulphonate(s)-(+)-oxirane-2-methanol p-toluenesulphonate

H56000-14 H56000-06 H56000-03
AAH56000-14EA 409.59 USD
AAH56000-14 AAH56000-03 AAH56000-06
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulfonate 99%
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate
Formula: C₁₀H₁₂O₄S
MW: 228.26 g/mol
Melting Pt: 46…49 °C
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064489
CAS Number: 70987-78-9
EINECS: 417-210-7
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR