4-Carboxy-2-fluorophenylboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-Carboxy-2-fluorophenylboronic Acid Pinacol Ester4-Carboxy-2-fluorobenzeneboronic acid pinacol ester

H62028-03 H62028-MD H62028-06
AAH62028-06EA 352.6 USD
AAH62028-06 AAH62028-MD AAH62028-03
4-Carboxy-2-fluorophenylboronic acid pinacol ester 96%
4-Carboxy-2-fluorophenylboronic acid pinacol ester
Formula: C₁₃H₁₆BFO₄
MW: 266.07 g/mol
Melting Pt: 199…204 °C
Storage Temperature: Ambient
MDL Number: MFCD12756466
CAS Number: 1050423-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR