(2-Bromo-5-fluorophenyl)acetic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-5-fluorophenylacetic acid

H62005-03 H62005-14 H62005-06
AAH62005-06EA 729.99 USD
AAH62005-06 AAH62005-03 AAH62005-14
(2-Bromo-5-fluorophenyl)acetic acid 96%
(2-Bromo-5-fluorophenyl)acetic acid
Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
Melting Pt: 113…114 °C
Storage Temperature: Ambient
MDL Number: MFCD03094440
CAS Number: 739336-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR