2-Amino-6-bromobenzonitrile 95%

Warning

H61927-MD H61927-06 H61927-03
AAH61927-03EA 101.17 USD
AAH61927-03 AAH61927-MD AAH61927-06
2-Amino-6-bromobenzonitrile 95%
2-Amino-6-bromobenzonitrile
Formula: C₇H₅BrN₂
MW: 197.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834769
CAS Number: 77326-62-6
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR