2-Bromo-4-fluoro-6-nitroaniline tech. 90%, tech.

Warning

H61840-06 H61840-03
AAH61840-03EA 61.14 USD
AAH61840-03 AAH61840-06
2-Bromo-4-fluoro-6-nitroaniline tech. 90%, tech.
2-Bromo-4-fluoro-6-nitroaniline
Formula: C₆H₄BrFN₂O₂
MW: 235.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779528
CAS Number: 10472-88-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR