1-(Mercaptomethyl)cyclopropaneacetic acid 98%

Danger

Synonyms: 2-(1-(Mercaptomethyl)cyclopropyl)acetic acid[1-(Thiomethyl)cycloprop-1-yl]acetic acid

H61723-03 H61723-06
AAH61723-03EA 74.32 USD
AAH61723-03 AAH61723-06
1-(Mercaptomethyl)cyclopropaneacetic acid 98%
1-(Mercaptomethyl)cyclopropaneacetic acid
Formula: C₆H₁₀O₂S
MW: 146.20 g/mol
Melting Pt: 42…46 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03411600
CAS Number: 162515-68-6
EINECS: 420-240-3
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR