N-Methyl-N-phenyl-β-alanine ethyl ester 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Methyl-N-[2-(ethoxycarbonyl)ethyl]anilineEthyl 3-[methyl(phenyl)amino]propionate

H61615-14 H61615-06
AAH61615-14EA 145.99 USD
AAH61615-14 AAH61615-06
N-Methyl-N-phenyl-β-alanine ethyl ester 99%
N-Methyl-N-phenyl-beta-alanine ethyl ester
Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Boiling Pt: 98…100 °C/0.05 mm
Storage Temperature: Ambient
MDL Number: MFCD01249502
CAS Number: 2003-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR