3-Bromo-2-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

H61169-03 H61169-06 H61169-MD
AAH61169-03EA 150.38 USD
AAH61169-03 AAH61169-06 AAH61169-MD
3-Bromo-2-nitroaniline 98%
3-Bromo-2-nitroaniline
Formula: C₆H₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08063248
CAS Number: 7138-15-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR