2-Chloro-5-nitronicotinic acid 95%
|
Warning
|
H66927-06
H66927-03
AAH66927-06EA
237.54
USD
AAH66927-06
AAH66927-03
2-Chloro-5-nitronicotinic acid 95%
2-Chloro-5-nitronicotinic acid
Formula:
C₆H₃ClN₂O₄ MW: 202.55 g/mol Melting Pt: 140…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD06658402 CAS Number: 42959-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...