Z-Tyr(tBu)-OMe 95%

Synonyms: (S)-Methyl 2-(((benzyloxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoateN-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester

H66925-03 H66925-06 H66925-14
AAH66925-06EA 103.56 USD
AAH66925-06 AAH66925-03 AAH66925-14
Z-Tyr(tBu)-OMe 95%
Z-Tyr(tBu)-OMe
Formula: C₂₂H₂₇NO₅
MW: 385.46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237370
CAS Number: 5068-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR