(S)-1-Benzyl-2-(2,5-dioxopyrrolidin-1-yl)-5-oxopyrrolidine-1,2-dicarboxylate 95%

Synonyms: Z-Pyr-OSu

H66858-06 H66858-14 H66858-03
AAH66858-14EA 425.13 USD
AAH66858-14 AAH66858-03 AAH66858-06
(S)-1-Benzyl-2-(2,5-dioxopyrrolidin-1-yl)-5-oxopyrrolidine-1,2-dicarboxylate 95%
(S)-1-Benzyl-2-(2,5-dioxopyrrolidin-1-yl)-5-oxopyrrolidine-1,2-dicarboxylate
Formula: C₁₇H₁₆N₂O₇
MW: 360.322 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00191144
CAS Number: 40291-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR