Ac-D-Pro-OH 98%

Synonyms: N-Acetyl-D-proline

H66524-14 H66524-06
AAH66524-14EA 180.93 USD
AAH66524-14 AAH66524-06
Ac-D-Pro-OH 98%
Ac-D-Pro-OH
Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00080841
CAS Number: 59785-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR