Boc-D-Glu(Ochex)-OH 95%

Synonyms: N-Boc-D-glutamic acid 5-cyclohexyl ester

H66472-MD H66472-03 H66472-06
AAH66472-03EA 66.44 USD
AAH66472-03 AAH66472-06 AAH66472-MD
Boc-D-Glu(Ochex)-OH 95%
Boc-D-Glu(Ochex)-OH
Formula: C₁₆H₂₇NO₆
Storage Temperature: Freezer
MDL Number: MFCD00076929
CAS Number: 133464-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR