Z-Arg(Mbs)-OH·DCHA 95%

Synonyms: Z-Arg(Mbs)-OH·DCHA

H66240-06 H66240-03 H66240-MD
AAH66240-03EA 69.2 USD
AAH66240-03 AAH66240-MD AAH66240-06
Z-Arg(Mbs)-OH·DCHA 95%
Z-Arg(Mbs)-OH·DCHA
Formula: C₃₃H₄₉N₅O₇S
MW: 659.843 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237326
CAS Number: 58810-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR