2-Amino-3-bromo-6-methylpyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-3-bromo-6-picoline 3-Bromo-6-methyl-2-pyridylamine 3-Bromo-6-methylpyridin-2-amine

H66162-06 H66162-03
AAH66162-06EA 183.99 USD
AAH66162-06 AAH66162-03
2-Amino-3-bromo-6-methylpyridine 95%
2-Amino-3-bromo-6-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 78…83 °C
Storage Temperature: Ambient
MDL Number: MFCD03095215
CAS Number: 126325-46-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR