N-Acetyl-D-valine 98%

Warning

Synonyms: Ac-D-Val-OH

H66103-14 H66103-22 H66103-06
AAH66103-22EA 445.11 USD
AAH66103-22 AAH66103-14 AAH66103-06
N-Acetyl-D-valine 98%
N-Acetyl-D-valine
Formula: C₇H₁₃NO₃
MW: 159.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03939895
CAS Number: 17916-88-0
EINECS: 241-860-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR