Boc-DL-Tle-Oh 96%

Synonyms: Boc-tBu-DL-Gly-OHN-Boc-DL-tert-leucine

H66033-MD H66033-03 H66033-06
AAH66033-MDEA 23.23 USD
AAH66033-MD AAH66033-03 AAH66033-06
Boc-DL-Tle-Oh 96%
Boc-DL-Tle-Oh
Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057782
CAS Number: 102185-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR