(S)-Methyl-2-aminohexanoate hydrochloride 98%

Synonyms: H-Nle-OMe·HCl

H65873-14 H65873-03 H65873-06
AAH65873-06EA 116.63 USD
AAH65873-06 AAH65873-14 AAH65873-03
(S)-Methyl-2-aminohexanoate hydrochloride 98%
(S)-Methyl-2-aminohexanoate hydrochloride
Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077151
CAS Number: 3844-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR