N(α)-Benzyloxycarbonyl-D-lysine 95%

Synonyms: Z-D-Lys-OH

H65640-03 H65640-06
AAH65640-06EA 242.56 USD
AAH65640-06 AAH65640-03
N(α)-Benzyloxycarbonyl-D-lysine 95%
N(α)-Benzyloxycarbonyl-D-lysine
Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067713
CAS Number: 70671-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR