H-Met-Otbu hydrochloride 95%

Synonyms: H-Met-OtBu·HCl

H65439-03 H65439-MD H65439-06
AAH65439-MDEA 43.09 USD
AAH65439-MD AAH65439-06 AAH65439-03
H-Met-Otbu hydrochloride 95%
H-Met-Otbu hydrochloride
Formula: C₉H₂₀ClNO₂S
MW: 241.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038896
CAS Number: 91183-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR