Z-Ser-Gly-OET 95%

Synonyms: Z-Ser-Gly-OEt

H65328-03 H65328-06 H65328-MD
AAH65328-06EA 407.37 USD
AAH65328-06 AAH65328-MD AAH65328-03
Z-Ser-Gly-OET 95%
Z-Ser-Gly-OET
Formula: C₁₅H₂₀N₂O₆
MW: 324.333 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191150
CAS Number: 4526-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR