Z-D-Asp(OBzl)-OH 95%

Synonyms: Z-D-Asp(OBzl)N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester

H65325-03 H65325-MD H65325-06
AAH65325-03EA 84.85 USD
AAH65325-03 AAH65325-06 AAH65325-MD
Z-D-Asp(OBzl)-OH 95%
Z-D-Asp(OBzl)-OH
Formula: C₁₉H₁₉NO₆
MW: 357.362 g/mol
Melting Pt: 105…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00237336
CAS Number: 5241-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR