1-Benzhydrylazetidine-3-carbonitrile 97%

Warning

H64647-06 H64647-14
AAH64647-06EA 343.74 USD
AAH64647-06 AAH64647-14
1-Benzhydrylazetidine-3-carbonitrile 97%
1-Benzhydrylazetidine-3-carbonitrile
Formula: C₁₇H₁₆N₂
MW: 248.329 g/mol
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00158910
CAS Number: 36476-86-5
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR