2,6-Dichloro-3-iodopyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H64366-03 H64366-06
AAH64366-03EA 31.72 USD
AAH64366-03 AAH64366-06
2,6-Dichloro-3-iodopyridine 98%
2,6-Dichloro-3-iodopyridine
Formula: C₅H₂Cl₂IN
MW: 273.88 g/mol
Melting Pt: 74…79 °C
Storage Temperature: Ambient
MDL Number: MFCD03094686
CAS Number: 148493-37-2
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR