1-Boc-6-bromoindole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: tert-Butyl 6-bromo-1H-indole-1-carboxylate

H63538-MD H63538-06 H63538-03
AAH63538-MDEA 42.89 USD
AAH63538-MD AAH63538-06 AAH63538-03
1-Boc-6-bromoindole 97%
1-Boc-6-bromoindole
Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954826
CAS Number: 147621-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR