4-Chloro-1H-indazole 97%

Danger

H37416-06 H37416-03
AAH37416-03EA 126.17 USD
AAH37416-03 AAH37416-06
4-Chloro-1H-indazole 97%
4-Chloro-1H-indazole
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00211065
CAS Number: 13096-96-3
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR