Manganesebis(trifluoromethylsulfonyl)imide

Danger

Synonyms: Manganese bis(trifluoromethanesulfonyl)imide

47179-MD 47179-03 47179-06
AA47179-03EA 115.44 USD
AA47179-03 AA47179-06 AA47179-MD
Manganesebis(trifluoromethylsulfonyl)imide
Manganesebis(trifluoromethylsulfonyl)imide
Formula: C₄F₁₂MnN₂O₈S₄
MW: 615.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380173
CAS Number: 207861-55-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR