Mercurybis(trifluoromethylsulfonyl)imide

Danger

Synonyms: Mercury bis(trifluoromethanesulfonyl)imideMercury bis(trifluoromethanesulfonimide)

47156-MD 47156-06
AA47156-MDEA 36.95 USD
AA47156-MD AA47156-06
Mercurybis(trifluoromethylsulfonyl)imide
Mercurybis(trifluoromethylsulfonyl)imide
Formula: C₄F₁₂HgN₂O₈S₄
MW: 760.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380172
CAS Number: 104715-41-5
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR