Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "ozone-generators"
Ozone generators are designed to produce a continuous flow of ozone. The ozone gas is designed for calibration of field and laboratory ozone analyzers in water purification and air pollution applications. It utilizes the photochemical reaction of oxygen under shortwave UV to produce a continuous flow of ozone. Generators are a stable source for instrument development and an oxidant source for aging and oxidant testing of materials. Low-maintenance systems eliminate inconvenient extended downtime.
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Analytik Jena US
Description:   Ozone generators produce a continuous flow of ozone as well as a stable source for water purification and air pollution studies. Designed for calibrat...
Product available on GSA Advantage®