Would you like to log in again?
Cell Migration Assay, Flex Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT117-0001
Reproducibly monitor real-time cell migration, without the use of special instrumentation.
2D assay for investigating cell migration of adherent cell lines.
The Cell migration assay, flex kit is a reproducible, sensitive, and flexible assay that can be used to monitor cell migration or cell invasion. The Cell migration assay, flex kit now gives researchers more control over designing a cell migration assay. Each Cell migration assy, flex kit contains the Cell Seeding Stoppers for creating a detection zone at the center of each well in a 96-well plate. Since the stoppers are not pre-inserted into the wells, researchers can coat the plate with an extracellular matrix (ECM) component to design their assay. Researchers may also apply 3-dimensional overlays in each well to watch how cells invade and respond to various compounds (chemokinesis).The Cell migration assay, flex kit is designed to be used with any stain or labeling technique. Readout can be performed by microscopy or use of a microplate reader.The Cell migration assay, flex kit has been designed for use with adherent cell cultures. This assay has been successfully used with 3T3-Swiss albino, HT-1080, HCEC, HUVEC, and MCF10A cell lines.
Ordering information: Contains tissue culture treated, stopper compatible 96-well plate (black, clear bottom), cell seeding stoppers (96), cell migration detection mask, cell migration stopper tool.
ORDER
Expand All / Collapse All
No. of Wells Supplier No. VWR Catalog Number Unit Price Quantity
96 KIT117-0001 10014-614 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty