Motic BA210 Upright Compound Microscope, Motic Instruments
1100100401452
76164-554EA 1781.18 USD
76164-554
Motic BA210 Upright Compound Microscope, Motic Instruments
The Motic BA210 is a versatile microscope intended for both educational and professional environments.
  • LED illumination 3W with intensity control
  • Vertical travel range 25 mm
The BA210 features a Siedentopf style head inclined at 30° with full 360° rotation for comfort while viewing for extended periods of time. Coaxial coarse and fine focusing adjustment, built-in low position coaxial mechanical stage, and a focusable Abbe condenser N.A. 1.25 with iris diaphragm and slot help ensure professional, repeatable image quality for all intended applications. The BA210 is a robust student instrument that brings professional, repeatable image quality results to all of its intended applications.
With the addition of Motic’s CCIS Infinity Optics, the BA210 provides exceptional image quality with perfect flatness of field. Available add-ons for applications such as phase contrast, darkfield, or polarization microscopy transform the BA210 into a truly robust instrument for both educational and industrial purposes.
Ordering information: Microscope is supplied with all accessories including immersion oil 5 ml, power cord, cord hanger, dustcover, allen key, thumb screw and spare fused. Contact your VWR representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.