LED Transilluminator, Dual Wavelength, G-Biosciences
BT508
10001-858EA 1566.01 USD
10001-858
LED Transilluminator, Dual Wavelength, G-Biosciences
The Dual wavelength LED Transilluminator has high uniformity across the whole viewing area.
Application (In-Cell westerns) as an excitation source with strong R/NIR LEDs for excitation. Dual wavelengths are embedded in the same transilluminator. Delta LED arrangement for 50,000 hours.
Weight: 3.6 kg
Certifications: US Patent Pending. CE/ FCC certified.