Jackson Safety* H20 Reusable Triple Flange Earplugs - Uncorded, KIMBERLY-CLARK PROFESSIONAL®
67220
10127-394CS 509.77 USD
10127-394
Jackson Safety* H20 Reusable Triple Flange Earplugs - Uncorded, KIMBERLY-CLARK PROFESSIONAL®
JACKSON SAFETY® H20 Reusable Triple Flange Earplugs feature class A(L) washable ridged stems for better grip and easier insertion and removal.
  • Class A(L) washable ridged stems for better grip and easier insertion/removal
  • Conforms to ear canal for comfortable fit and seal
  • ANSI S3.19
Three soft flanges collapse to conform to the ear canal for a comfortable fit and seal.
NRR: 26
Certifications: ANSI S3.19-1974.
Ordering information: Individually packaged for hygienic storage.