Duo-Check Combined Class 5/Biological Indicator Process Challenge Pack, Propper
26940500
10798-792CS 259.01 USD
10798-792
Duo-Check Combined Class 5/Biological Indicator Process Challenge Pack, Propper
The Duo-Check Combined Challenge Pack includes a Type 5 integrator and a self-contained biological indicator
It is recommended for the monitoring and release of every steam sterilizer load. Ideal for immediate load release in emergency situations using integrator results backed by the biological indicator.