Would you like to log in again?
Vapor Line Class 5 Integrating Indicators, Propper
Supplier: PROPPER MFG CO INC
26900510 26900825
The Vapor Line Type 5 Integrator is a chemical indicator designed for use in monitoring saturated steam, temperature, and time in steam sterilizers
The integrator is equivalent in performance to biological indicators. A PASS result assures that the conditions for sterilization inside the pack were achieved.
Certifications: AAMI/ISO 11140-1:2014, Type 5
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Vapor Line Type 5 Integrating Indicators 26900510 10798-786 Pack of 100 Retrieving Restricted
Product and/or packaging is safer to human and environmental health
Vapor Line Type 5 Integrating Indicators 26900825 10798-788 Pack of 250 Retrieving Restricted
Product and/or packaging is safer to human and environmental health
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging is safer to human and environmental health
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty