Motic B1-Series Upright Microscope, Motic Instruments
1100100500333 1100100500333 1100100500111 1100100500201 1100100500201 1100100500261 1101001200091 1101001200091 1100400800011 1100100500111
19000-790EA 965.94 USD
19000-790 CA19000-790 19000-792 19000-168 19000-850 CA19000-168 CA19000-850 19000-822 19000-170 CA19000-170
Motic B1-Series Upright Microscope, Motic Instruments
Motic’s B1 Series Microscope range provides users a sturdy base for educational or light-duty laboratory applications
The B1 Compound Microscope features a 45° inclined binocular head that can rotate 360° for comfortable viewing. The B1 series microscopes come standard with 4x, 10x, 40x S, and 100x S-oil Achromatic objectives. These can be upgraded to achromatic super contrast (ASC) objectives for additional contrast. Superb optical clarity and reliable mechanical engineering assure years of top performance.
The B1 series is ideal for beginner and novice users, with large focusing knobs and stage controls to provide easy operation while the widefield eyepieces are more forgiving of the user’s head and eye movement. Coaxial stage controls and illumination dimmer knobs are conveniently located to enable uninterrupted "eyes-down" operation. A wide range of accessories are available to meet most microscopy applications including phase contrast and darkfield attachments.
Ordering information: Microscope is supplied with micrometer reticle, polarizing equipment, phase contrast accessories, Darkfield accessories, and objective upgrades. Contact your VWR representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.