Emu-Chex™ Emulating Indicator Strips, Propper Manufacturing
26611600
10798-530PK 30.95 USD
10798-530
Emu-Chex™ Emulating Indicator Strips, Propper Manufacturing
Emu-Chex™ type 6 indicator strips monitor all critical variables of specific sterilization cycles.
  • Type 6 chemical indicator
  • Non-toxic heavy metal free ink formulation
  • Distinct green to red color change that will not revert
  • Can be used as a permanent record
These indicator strips provide a reliable visual confirmation that sterilization conditions have been met, when used correctly. It features circular indicator mark that will change from green to red when ideal sterilization parameters have been met for your chosen cycle.
Certifications: AAMI/ISO 11140-1:2005 standards, for Type 6 chemical indicators.