Would you like to log in again?
Easy Piercing Seal, BioChromato
Supplier: BioChromato
R37.001.00
102962-948BX 207.9 USD InStock
102962-948
Easy Piercing Seal, BioChromato
Seals Microplate Sealing Films and Foils
Easy Piercing Seal (EPS) punctures easily to prevent needle clogging
  • Punctures easily
  • Prevents needle clogging
  • No adhesive residues are left on the plate
  • No adhesive residues are left on the injector needle
  • Highly resistant to orgain solvents
In addition, no adhesive residues are left on the plate or injector needle. The EPS plate seal is made of polyolefin and is highly resistant to organic solvents. Suggested applications include HPLC and LC/MS.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty